missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA Clean-up XS Column
£53.95 - £1090.00
Specifications
Detection Method | One-phase extraction |
---|---|
Isolation Technology | One-phase Extraction |
Final Product Type | NucleoSpin™ RNA Clean-up XS Column |
For Use With (Application) | For RNA isolation from cultured cells, animal tissue, plant tissue, microorganisms |
Includes | Collection Tubes, Collection Tubes (2 mL), Collection Tubes (1.5 mL), buffers |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11912482
|
Macherey-Nagel™
740903.10 |
10 |
£53.95
10 reactions |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12743580
|
Macherey-Nagel™
740903.250 |
250 |
£1090.00
250 reactions |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12753580
|
Macherey-Nagel™
740903.50 |
50 |
£381.00
50 reactions |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
One-phase extraction | |
NucleoSpin™ RNA Clean-up XS Column | |
Collection Tubes, Collection Tubes (2 mL), Collection Tubes (1.5 mL), buffers | |
1 Each |
One-phase Extraction | |
For RNA isolation from cultured cells, animal tissue, plant tissue, microorganisms | |
NucleoSpin™ RNA Clean-up XS Column | |
Reagent |