Learn More
Macherey-Nagel™ NucleoSpin™ RNA Clean-up Maxi
Complete kits for simple, fast, and convenient clean up of huge RNA amounts (pre-purified RNA, reaction mixtures, DNase reaction mixtures).
Brand: Macherey-Nagel™ 740910.20
Additional Details : Weight : 0.40000kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
- Designed to clean up RNA from either crude preparations or enzyme reactions
- Designed for very large samples – up to 35 mg crude RNA
- Complete removal of RT-PCR inhibitors
- Time saving procedure without DNase digestion
Specifications
Room temperature (18-25 °C) | |
Silica Membrane | |
Crude RNA | |
Ultra-pure RNA | |
Clean-up of large amounts of RNA | |
20 | |
Up to 35 mg | |
85 - 95% |
Maxi Spin Column | |
3 - 5 mL | |
Binding capacity: 35 mg | |
Vortex Mixer, Centrifuge for 50 mL Tubes | |
NucleoSpin RNA Clean-up Maxi Columns, Collection Tubes, buffers | |
A260/A280: 1.9 - 2.1 | |
30 min/6 preps |
Safety and Handling
- NucleoSpin RNA Clean-up Maxi (20)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : guanidium thiocyanide, ethanol