missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Clean-up, Mini kit for RNA clean up and concentration

Product Code. 12708612
Change view
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
10 reactions
250 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12708612 50 50 reactions
11992482 10 10 reactions
12798602 250 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12708612 Supplier Macherey-Nagel™ Supplier No. 740948.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recommended for the clean-up of total RNA from RNA preparations which contain inacceptable amounts of RT-PCR* inhibitors.

NucleoSpin™ RNA clean-up kits are recommended for the clean-up of total RNA from RNA preparations which contain inacceptable amounts of RT-PCR* inhibitors, often found in e.g., RNA prepared with phenol-chloroform based methods. It is further recommended for the isolation of RNA from small amounts of cultured cells whenever copurification of some genomic DNA is acceptable. For the isolation of RNA from cells and tissue with lowest DNA contamination of the isolated RNA we recommend DNase I containing NucleoSpin™ RNA kits (see ordering information). The kits allow purification of pure RNA with an A260/280 ratio generally exceeding 1.9 (measured in TE buffer (pH7.5)). NucleoSpin™ RNA clean-up kits are further recommended for the clean-up of RNA from enzymatic reactions like in vitro transcribed RNA, amplification reactions (e.g., ExpressArt™ Amino-Allyl-mRNA Amplification Kit, artus GmbH, Hamburg, Germany), biotinylated RNA or fluorescent (Cy dye) labelled RNA. The purified RNA is ready to use for applications like enzymatic labelling reactions (e.g., dye incorporation), reverse transcriptase-PCR (RT-PCR), and for DNA/RNA based chip hybridisations (e.g., MWG rat microarray, MWG, Ebersberg, Germany or Human Genome U133A Array, Affymetrix, USA). Integrity of purified RNA, originally isolated from e.g., eukaryotic cells, is examined by denaturing agarose gel electrophoresis: rRNA bands are sharp, with the 28S band being about twice as intense as the 18S band.
  • Silica membrane technology
  • Typical recovery: 85% to 95%
  • Sample material: ≤200μL phenol/chloroform extract or reaction mixture
  • Elution volume: 40μL to 100μL
  • Fragment size: >200b
  • Binding capacity: 200μg
  • Preparation time: 20 min./6 preps
  • Format: mini spin column
  • RNA suitable for RT-PCR, Northern blotting, RNase protection assays, primer extension and array technology For RNA clean up of pre-purified RNA (e.g., Trizol); Reaction mixtures; RNA from small cell numbers; Amino-Allyl-mRNA; Biotinylated RNA.

Specifications

Detection Method One-phase extraction
Isolation Technology One-phase Extraction
Final Product Type NucleoSpin™ RNA Clean-up Column
For Use With (Application) For RNA isolation from cultured cells, animal tissue, plant tissue, microorganisms
Includes Collection Tubes, Collection Tubes (2 mL), Collection Tubes (1.5 mL), buffers
No. of Reactions 50
Product Type NucleoSpin™ RNA Clean-up Column
Quantity 1 Each
Sample Size <200 μL
Form Reagent
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin RNA Clean-up
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate, ethanol (denatured with 1%IPA/1%MEK, acc.2016/1867/EU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.