missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Blood, Mini kit for RNA from blood

Product Code. p-8013754
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
120mL
This item is not returnable. View return policy

Product Code. 12825693

Brand: Macherey-Nagel™ 740200.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate RNA with a simple and convenient spin column format. Macherey Nagel™ NucleoSpin™ RNA Blood is optimized for isolation of RNA from fresh or frozen whole blood (human or animal).

Features
  • Direct total blood lysis – very simple and convenient procedure (patent pending)
  • No selective erythrocyte lysis at 4 °C – complete processing at room temperature
  • Superior RNA yield and quality from up to 400 μL whole blood
  • Efficient on-column DNA removal for an increased sensitivity in downstream applications Compatible with common blood collection tubes and anticoagulants
  • (e.g., EDTA, citrate, and heparin)
Applications
  • Isolation of RNA from fresh or frozen whole blood from common blood collection tubes
  • (EDTA, citrate, heparin)
  • Typical downstream applications: qRT-PCR, next generation sequencing, blotting, array technology

Specifications

Format Mini Spin Column
For Use With (Application) Isolation of RNA from fresh or frozen whole blood from common blood collection tubes
Includes NucleoSpin™ RNA Blood Columns with Collection Tubes, Collection Tubes (2 mL), buffers, RNase-free rDNase, Liquid Proteinase K
No. of Reactions 10
Product Type NucleoSpin RNA Blood
RNase Yes
Sample Size <400 μL
Test Time 55 min./6 Reaction
Yield 1 to 8μg
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate,ethanol (diluted< 20 %) /guanidinehydrochloride,ethanol/
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.