missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ RNA Blood, Mini kit for RNA from blood
Description
Features
- Direct total blood lysis – very simple and convenient procedure (patent pending)
- No selective erythrocyte lysis at 4 °C – complete processing at room temperature
- Superior RNA yield and quality from up to 400 μL whole blood
- Efficient on-column DNA removal for an increased sensitivity in downstream applications Compatible with common blood collection tubes and anticoagulants
- (e.g., EDTA, citrate, and heparin)
- Isolation of RNA from fresh or frozen whole blood from common blood collection tubes
- (EDTA, citrate, heparin)
- Typical downstream applications: qRT-PCR, next generation sequencing, blotting, array technology
Specifications
Specifications
| Format | Mini Spin Column |
| For Use With (Application) | Isolation of RNA from fresh or frozen whole blood from common blood collection tubes |
| Includes | NucleoSpin™ RNA Blood Columns with Collection Tubes, Collection Tubes (2 mL), buffers, RNase-free rDNase, Liquid Proteinase K |
| No. of Reactions | 10 |
| Product Type | NucleoSpin RNA Blood |
| RNase | Yes |
| Sample Size | <400 μL |
| Test Time | 55 min./6 Reaction |
| Yield | 1 to 8μg |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: guanidinium thiocyanate,ethanol (diluted< 20 %) /guanidinehydrochloride,ethanol/
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?