missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Blood Midi

Product Code. 15128584
Change view
Click to view available options
Format:
Midi Spin Column
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
15128584 Midi Spin Column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15128584 Supplier Macherey-Nagel™ Supplier No. 740210.20
  • ONLINE EXCLUSIVE PRICE
    £280.88
    £255.73 / Pack of 1
    Save £25.15
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Midi spin kit for superior isolation of total RNA from fresh or frozen whole blood (human or animal) from common blood collection tubes. Direct blood lysis process does not require the use of a selective erythrocyte lysis.

NucleoSpin™ RNA Blood Midi kit achieves superior RNA yield and quality in a format that provides convenient handling and flexible sample volume and throughput.

  • Direct total blood lysis—Very simple and convenient procedure
  • No selective erythrocyte lysis at 4 °C—Complete processing at room temperature
  • Superior RNA yield and quality from up to 1.3 mL whole blood
  • Efficient on-column DNA removal for an increased sensitivity in downstream applications
  • Compatible with common blood collection tubes and anticoagulants (e.g., EDTA, citrate, and heparin)

Specifications

Format Midi Spin Column
Isolation Technology Silica Membrane Technology
Elution Volume 200 to 400 μL
Sample Type Whole Blood (Fresh or Frozen)
Efficiency Binding capacity: 700 μg
Final Product Type Total RNA
For Use With (Equipment) Vortex Mixer, Centrifuge
For Use With (Application) RNA isolation
Includes NucleoSpin RNA Blood Midi Columns with Collection Tubes, Collection Tubes (15 mL), buffers, RNase-free rDNase, Liquid Proteinase K
No. of Reactions 20 Preps
Purity A260/A280: 1.9 - 2.1
Sample Size 400 to 1300 μL
Test Time 75 min/6 Preps
Yield 4 to 26 μL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin RNA Blood Midi (20)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate; ethanol (diluted < 20 %) (denaturedwith 1%IPA/1%MEK, acc.2016/1867/EU); pr
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.