missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin Plasmid QuickPure™ Kit

Product Code. 11902422
Change view
Click to view available options
No. of Reactions:
250 Preps
50 Preps
Unit Size:
250 reactions
50 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
11902422 250 Preps 250 reactions
11902362 50 Preps 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11902422 Supplier Macherey-Nagel™ Supplier No. 740615.250

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Features a specially treated silica membrane that speeds up the procedure by combining washing and drying into one step. Macherey-Nagel™ NucleoSpin Plasmid QuickPure™ provides ultra-fast small-scale plasmid DNA purification which offers a saving of time and costs.

With the NucleoSpin™ Plasmid QuickPure method, the bacteria culture is lysed according to alkaline lysis. The lysate is cleared by centrifugation and the supernatant is loaded onto the NucleoSpin™ Plasmid QuickPure column. Contaminations like salts, metabolites, nucleases and soluble macromolecular cellular components are removed by only one single washing step with buffer AQ. Pure plasmid DNA is finally eluted under low ionic strength conditions with slightly alkaline buffer AE (5mM Tris-Cl, pH 8.5). The prepared plasmid DNA is suitable for all common downstream applications.

Kit components:
Resuspension Buffer, Lysis Buffer, Neutralization Buffer, Wash Buffer, Elution Buffer, RNase A, NucleoSpin™ Plasmid QuickPure Columns, Collection Tubes, User manual

Features:

  • Yield: up to 15μg
  • Ultra-fast procedure: 18 minipreps in 11min
  • Elution volume: 50μL
  • Sample material: 1mL to 3mL E. coli culture
  • Very good price-performance ratio
  • Silica-membrane technology
  • Binding capacity: 15μg
  • No use of organic solvents
  • Ultra-fast small scale preparation of plasmid DNA
  • Highly pure nucleic acids suitable for all common downstream applications
  • Vectors <15kb
  • Format: mini-spin columns
For further information on this product please contact Customer Services.

Specifications

Content And Storage Room temperature (18–25°C)
Format Mini Spin Column
For Use With (Equipment) Standard benchtop microcentrifuge
For Use With (Application) Plasmid DNA purification/isolation
Includes Resuspension Buffer A1, Lysis Buffer A2 (without LyseControl), Neutralization Buffer A3, Wash Buffer AQ (Concentrate), Elution Buffer AE, RNase A (lyophilized), NucleoSpin™ Plasmid QuickPure Columns (dark yellow rings), Collection Tubes (2mL), User manual
No. of Reactions 250 Preps
Product Type NucleoSpin Plasmid QuickPure
RNase Lyophilized
Sample Volume Culture volume: 1–3mL high copy
Size <15 kbp
Test Time Prep time: 11min./18 preps
Yield <15μg (1–3mL culture)
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.