missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid QuickPure Columns

Product Code. 15810938
Change view
Click to view available options
Quantity:
250 Each
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15810938 250 Each Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15810938 Supplier Macherey-Nagel™ Supplier No. 740615.250S

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mini spin columns for use with NucleoSpin™ Plasmid QuickPure kits feature a specially treated silica membrane to speed up the preparation of high-purity plasmid DNA from E. coli culture through a combined washing and drying step.

NucleoSpin™ Plasmid EasyPure

  • Utra-fast small scale preparation high-purity plasmid DNA: 6 mini preps in just 11 min
  • Contaminations are removed by a single washing-drying step
  • Downstream applications: cloning, sequencing, PCR, transformation, restriction analysis
    • High-copy plasmids from E. coli
    • Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required)
    • Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures
    • Supplementary protocol for M13 DNA availabl

Specifications

Color Clear
Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Sample Type E. coli Culture
Efficiency Binding capacity: 15 μg
Elution Volume Elution volume: 50 μL
Final Product Type Sequencing-grade plasmid DNA
For Use With (Equipment) NucleoSpin™ Plasmid QuickPure Kit
For Use With (Application) Preparation of high-purity plasmid DNA
Includes Columns only
Product Type QuickPure Column
Quantity 250 Each
Sample Volume 1 - 3 mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin Plasmid QuickPure
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.