missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid (NoLid) Columns for plasmid DNA purification

Product Code. p-7159605
Click to view available options
Quantity:
250 Pk.
50 Pk.
Unit Size:
Pack of 250
Pack of 50
This item is not returnable. View return policy

Product Code. 15860918

Brand: Macherey-Nagel™ 740499.50S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Replacement columns for use with NucleoSpin™ Plasmid (NoLid) kits for rapid, small-scale preparation of high-purity, sequencing-grade plasmid DNA.

NucleoSpin™ Plasmid (NoLid)

  • For rapid small scale preparation high-purity plasmid DNA
  • Fast procedure – 6 Mini preps in 25 min
  • High capacity – up to 50 μg of plasmid DNA
  • Chaotropic salt wash buffer – extra pure DNA
  • Downstream applications: cloning, sequencing, PCR, transformation, restriction analysis
    • High-copy plasmids from E. coli
    • Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required)
    • Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures
    • Supplementary protocol for M13 DNA availabl

Specifications

Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Sample Type E. coli Culture
Efficiency Binding capacity: 50 μg
Elution Volume 50 μL
Final Product Type Sequencing-grade plasmid DNA
For Use With (Equipment) NucleoSpin™ Plasmid (NoLid) Kit
For Use With (Application) Preparation of high-purity plasmid DNA
Includes Columns only
Product Type NucleoSpin Plasmid (NoLid) Column
Quantity 50 Pk.
Sample Volume 2 to 10 mL
Show More Show Less
Product Identifier
  • NucleoSpin Plasmid (NoLid)
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %); guanidine hydrochloride; 2-propanol;Rnase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.