missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Plant II Columns
Click to view available options
Quantity:
250 Columns
missing translation for 'unitSize'
Pack of 250
Beschreibung
NucleoSpin™ Plant II
- For rapid isolation of genomic DNA from plant samples with higher yield and quality
- Two alternative lysis buffers included for optimal processing of various samples and highest flexibility:
- Lysis Buffer PL1, based on CTAB lysis method
- Lysis Buffer PL2, based on SDS lysis method
- NucleoSpin™ Filters included for clarification of lysate
- Improved buffer compositions – higher yield and purity
- Optimized silica membrane – improved DNA binding
- RNase A included
Spezifikation
Spezifikation
| Binding Capacity | 50 μg |
| Kit Contents | Lid |
| Material | Silica membrane |
| Content And Storage | Room Temperature |
| Particle Size | 50 to ∼ 50 kbp |
| Quantity | 250 Columns |
| For Use With (Application) | Isolation of genomic DNA |
| Type | Spin Column |
| Sufficient For | Elution volume: 50 - 100 μL |
Sicherheit und Handhabung
Produktidentifikator
- NucleoSpin Plant II
- MIXTURE LIST-Contains : guanidine hydrochloride, ethanol/dodecylsulfate, sodium salt/guanidine hydrochloride, 2-propanol
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur