missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpinPlant II Buffers
Click to view available options
Quantity:
200 mL
1000 mL
Chemical Name or Material:
Binding Buffer PC
Elution Buffer PE
Lysis Buffer PL1
Lysis Buffer PL2
Precipitation Buffer PL3
Wash Buffer PW1
Wash Buffer PW2 Concentrate
Unit Size:
1000mL
200mL
Description
NucleoSpin™ Plant II Kits (mini, midi or maxi) are used for rapid preparation of genomic DNA from plant cells and fungi. Kits are available in capacities to accommodate small, medium and large scale processing.
- Two alternative lysis buffers included for optimal processing of various samples and highest flexibility:
- Lysis Buffer PL1, based on CTAB lysis method
- Lysis Buffer PL2, based on SDS lysis method
- NucleoSpin™ filters included for clarification of lysate
- Improved buffer compositions – higher yield and purity
- Optimized silica membrane – improved DNA binding
- RNase A included
- Typical downstream applications: PCR, Southern blotting, enzymatic reactions
Specifications
Specifications
| Chemical Name or Material | Binding Buffer PC |
| CAS | 50-01-1 |
| Color | Yellow |
| For Use With (Application) | Preparation of genomic DNA from plant cells and fungi |
| For Use With (Equipment) | NucleoSpin Plant II kit |
| pH | 5.0 to 7.0 |
| Physical Form | Liquid |
| Quantity | 1000 mL |
| Formulation | Guanidine hydrochloride; Ethanol |
| Odor | Alcoholic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction