missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Microbial DNA Columns
Description
The NucleoSpin™ Microbial DNA kit is designed for convenient, fast, and easy isolation of genomic DNA from microbial samples. Because microbial samples can be difficult to lyse (due to their strong complex cell wall structures), the kit replaces enzymatic lysis with mechanical disruption of cell wall structures utilizing NucleoSpin™ Bead Tubes.
- Suitable for a large variety of starting materials
- High yields of genomic DNA can be isolated
- NucleoSpin Bead Tubes Type B included – for efficient lysis, compatible with many disruption devices
- Fast and easy procedure – Liquid Proteinase K included, no additional enzyme required
- Typical downstream applications: PCR, real-time PCR, southern blotting, enzymatic reactions
Specifications
Specifications
| Color | Green Band |
| Format | Mini Spin Column |
| Isolation Technology | Silica Membrane Technology |
| Sample Type | Bacteria (Gram +/-), Fungi, Yeast |
| Efficiency | Binding capacity: 60 μg |
| Final Product Type | Genomic DNA |
| For Use With (Equipment) | Centrifuge, NucleoSpin Microbial DNA kit |
| For Use With (Application) | DNA isolation from microorganisms |
| Quantity | 50 Pk. |
| Sample Volume | 500 to 600 μL |
| Show More |
Safety and Handling
Product Identifier
- NucleoSpin Microbial DNA
- Warning
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash with water thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- MIXTURE LIST-Contain: guanidine hydrochloride; 2-propanol; proteinase K, liquid (origin: tritirachiumalbum); guanidinium
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?