missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Microbial DNA Mini kit for DNA from microorganisms
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
10 reactions
250 reactions
50 reactions
Description
The NucleoSpin™ Microbial DNA kit is designed for convenient, fast, and easy isolation of genomic DNA from microbial samples. Because microbial samples can be difficult to lyse (due to their strong complex cell wall structures), the kit replaces enzymatic lysis with mechanical disruption of cell wall structures utilizing NucleoSpin™ Bead Tubes.
- Suitable for a large variety of starting materials
- High yields of genomic DNA can be isolated
- NucleoSpin Bead Tubes Type B included – for efficient lysis, compatible with many disruption devices
- Fast and easy procedure – Liquid Proteinase K included, no additional enzyme required
- Typical downstream applications: PCR, real-time PCR, southern blotting, enzymatic reactions
Specifications
Specifications
| Format | Mini Spin Column |
| Isolation Technology | Silica MembraneTechnology and Mechanical (Bead Tube) Disruption |
| Sample Type | Bacteria (Gram +/-), Fungi, Yeast |
| Efficiency | Binding capacity: 60 μg |
| Elution Volume | Elution volume: 100 - 200 μL |
| Final Product Type | Genomic DNA |
| For Use With (Equipment) | Centrifuge, Vortex mixer |
| For Use With (Application) | DNA isolation from microorganisms |
| Includes | NucleoSpin Microbial DNA Columns, NucleoSpin Bead Tubes Type B, Collection Tubes (2 mL), buffers, Liquid Proteinase K |
| No. of Reactions | 250 |
| Show More |
Safety and Handling
Product Identifier
- NucleoSpin Microbial DNA
- Warning
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash with water thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- MIXTURE LIST-Contains : guanidine hydrochloride; 2-propanol; proteinase K, liquid (origin: tritirachiumalbum); guanidinium
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction