missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Microbial DNA Mini kit for DNA from microorganisms

Product Code. 15373609
Change view
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
10 reactions
250 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
15373609 50 50 reactions
15353609 10 10 reactions
15878096 250 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15373609 Supplier Macherey-Nagel™ Supplier No. 740235.50
  • ONLINE EXCLUSIVE PRICE
    £261.00
    £185.90 / 50 reactions
    Save £75.10
    28% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kit for reliable isolation of DNA from microorganisms (e.g. yeast, fungi, and gram-negative and gram-positive bacteria), with lysis that is supported by mechanical disruption with glass beads (tubes included).

The NucleoSpin™ Microbial DNA kit is designed for convenient, fast, and easy isolation of genomic DNA from microbial samples. Because microbial samples can be difficult to lyse (due to their strong complex cell wall structures), the kit replaces enzymatic lysis with mechanical disruption of cell wall structures utilizing NucleoSpin™ Bead Tubes.

  • Suitable for a large variety of starting materials
  • High yields of genomic DNA can be isolated
  • NucleoSpin Bead Tubes Type B included – for efficient lysis, compatible with many disruption devices
  • Fast and easy procedure – Liquid Proteinase K included, no additional enzyme required
  • Typical downstream applications: PCR, real-time PCR, southern blotting, enzymatic reactions

Specifications

Format Mini Spin Column
Isolation Technology Silica MembraneTechnology and Mechanical (Bead Tube) Disruption
Sample Type Bacteria (Gram +/-), Fungi, Yeast
Efficiency Binding capacity: 60 μg
Elution Volume Elution volume: 100 - 200 μL
Final Product Type Genomic DNA
For Use With (Equipment) Centrifuge, Vortex mixer
For Use With (Application) DNA isolation from microorganisms
Includes NucleoSpin Microbial DNA Columns, NucleoSpin Bead Tubes Type B, Collection Tubes (2 mL), buffers, Liquid Proteinase K
No. of Reactions 50
Purity A260/A280: 1.6 - 2.0
Sample Volume <40 mg wet weight
Test Time 35 min/prep
Yield 5 - 25 μg
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin Microbial DNA
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash with water thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: guanidine hydrochloride; 2-propanol; proteinase K, liquid (origin: tritirachiumalbum); guanidinium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.