missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up XS

Product Code. 17112806
Change view
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
17112806 50 Each
17192796 10 Each
17102806 250 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17112806 Supplier Macherey-Nagel™ Supplier No. 740611.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Concentrated clean up with small elution volumes from PCR reaction mixtures and agarose gels

  • Rapid concentration and clean up of PCR products
  • 2 in 1 kit for reaction mixtures and gel extraction
  • XS column design enables elution of highly concentrated DNA

Specifications

Capacity 5 μg
Column Type XS Spin Column
Content And Storage Non hazardous, 18°C to 25°C
Detection Method Silica Membrane Technology
Isolation Technology Silica Membrane Technology
Sample Type Agarose Gel, PCR Reaction Mixture
Elution Volume 6 to 12 μL
For Use With (Equipment) Centrifugation, Vacuum, Vortex Mixer
Gel Compatibility Agarose gel
For Use With (Application) PCR Clean-up and Gel Extraction, Sample Concentration
High-throughput Compatibility No
Includes NucleoSpin Gel and PCR Clean-up XS Columns, Collection Tubes, buffers
Indicator pH-Indicator included
Key Functions Gel extraction and PCR clean-up
No. of Reactions 50
Nucleic Acid Type DNA
Product Type Nucleic Acid Purification Kit
Purity A260/A280: 1.8-1.9
Sample Volume <200 μL PCR reaction mixture, <200 mg agarose gel
Shipping Condition Room Temperature
Size 50 bp-20 kbp
Target DNA Fragments
Test Time 15 min / 6 preps
Yield Recovery 70-95%
Show More Show Less
compliance-icons
Product Identifier
  • NucleoSpin Gel and PCR Clean-up XS (50)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidium thiocyanite

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.