missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ NucleoSpin™ Funnel Column

Product Code. 12798612
Change view
Click to view available options
Quantity:
30/Pk.
Unit Size:
30 sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12798612 30/Pk. 30 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12798612 Supplier Macherey-Nagel Bioanalysis™ Supplier No. 740959
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Set of NucleoSpin™ Funnel Column, Collection Tube (50 mL), Collection Tube (0.5 mL).

  • Funnel column
  • For use with NucleoSpin™ DNA Trace

Specifications

Content And Storage 18°C to 25°C
For Use With (Application) Isolation of DNA
Product Type Funnel column
Quantity 30/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.