missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ DNA Yeast for the isolation of genomic DNA from yeast

Product Code. 16855203
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
50 reactions
This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (2-Propanol mixture) Chem class 3 product group III

Product Code. 16855203

Brand: Macherey-Nagel™ 740236.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
UN 1993 Flammable liquid, n.o.s. (2-Propanol mixture) Chem class 3 product group III

Isolation of total DNA from yeast and fungi – MN Bead Tubes Type C included.

  • Optimized lysis with MN Bead Tubes Type C
  • Up to 100 mg of sample input
  • High genomic DNA yields

Specifications

Bacterial or Yeast Strain Yeast Strain
Cell Type Various yeast species including Saccharomyces cerevisiae, Schizosachharomyces pombe, and Pichia pastoris
Bead Type MN Bead Tubes Type C
Capacity 60 μg
Column Type Mini Spin Column
Content And Storage 18°C to 25°C
Description Isolation of total DNA from yeast and fungi
Detection Method Silica Membrane Technology
Format Mini Spin Column
Isolation Technology Silica MembraneTechnology and Mechanical (Bead Tube) Disruption
Sample Type Yeast Species including Saccharomyces cerevisiae, Schizosachharomyces pombe, and Pichia pastoris
Elution Volume 100 to 200 μL
Final Product Type Genomic DNA
For Use With (Equipment) Centrifuge, Vortex Mixer
For Use With (Application) Isolation of Total DNA From Yeast and Fungi, qRT-PCR
High-throughput Compatibility No
Includes MN Bead Tubes Type C, NucleoSpin DNA Yeast Columns, Collection Tubes, and Buffers
Key Functions Isolation of DNA
No. of Reactions 10
Nucleic Acid Type Genomic DNA
Product Type Nucleic Acid Purification Kit
Purity A260/A280: 1.6-2.0
Sample Volume <100 mg wet weight
Source Fungi, Yeast
Species Saccharomyces cerevisiae, Schizosachharomyces pombe, and Pichia pastoris
Target Genomic DNA
Technique Silica Membrane Technology
Test Time 35 min/prep
Yield Up to 20 μg
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin DNA Yeast
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 1-dodecylpyridiniumchloride, guanidiniumthiocyanate, proteinase K, guanidine hydrochloride, 2-propa

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.