missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ DNA RapidLyse, Mini kit for rapid DNA purification

Product Code. 15715422
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
10 reactions
250 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 15715422

Brand: Macherey-Nagel™ 740100.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Economical kit for the rapid isolation and purification of genomic DNA from tail and ear clippings, organs, and cells in one hour or less. Kits with 10, 50 or 250 preps available.

The isolation of genomic DNA (gDNA) can be a time-consuming task. The NucleoSpin™ DNA RapidLyse kit is designed with a unique buffer chemistry to minimize the expenditure of time, work, and money.

  • Unique lysis chemistry to efficiently release gDNA from cells, tissues, and organs
  • Powerful lysis in one hour or less
  • Superior gDNA yields compared to standard extraction methods
  • Highest cost-efficiency of gDNA purification products

Specifications

Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Sample Type Eukaryotic Cells, Organs (Fresh, Frozen, Dried and Ethanol-Preserved), Tail and Ear Clippings
Efficiency Binding capacity: 60 μg
Elution Volume Elution volume: 60 - 100 μL
Final Product Type genomic DNA
For Use With (Application) Rapid extraction of genomic DNA
Includes Columns, 2mL Collection tubes, Buffers, Poteinase K
No. of Reactions 50
Purity A260/A280: 1.7 - 1.9
Sample Volume Up to 40 mg fresh weight, 1 x 106 cells
Test Time 60 min or less
Yield Up to 4 μg DNA per mg tissue
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin DNA RapidLyse
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : proteinase K, liquid(origin: tritirachium album)/1-dodecylpyridiniumchloride,guanidinium thiocyanat
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.