missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ DNA Plasma Midi
Isolation of cell-free DNA from up to 5 mL blood plasma, utilizing the NucleoVac 96 Vacuum Manifold (sold separately, MPN 740681). The kit is designed for high recovery of circulating fragmented DNA ≥ 50 bp from human EDTA plasma.
£630.00 - £775.00
Specifications
Format | Midi Spin Column |
---|---|
Isolation Technology | Silica Membrane Technology |
Sample Type | Blood Plasma |
Elution Volume | Elution volume: 200 μL (140 μL final eluate) |
Final Product Type | Cell-free DNA |
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
15648455
|
Macherey-Nagel™
740302.48 |
Core Kit | NucleoSpin Plasma Midi Columns, Collection Tubes, buffers, Proteinase K |
£630.00
48 reactions |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15658455
|
Macherey-Nagel™
740303.48 |
Full Kit | NucleoSpin Plasma Midi Columns, Collection Tubes, buffers, Proteinase K, Lysis Blocks |
£775.00
48 reactions |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The NucleoSpin™ DNA Plasma Midi kit is designed for the efficient isolation of circulating DNA from human blood plasma. Fragmented DNA of 50 bp and larger can be purified with high efficiency.
- Superior recovery of fragmented cell-free DNA
- Parallel purification of 24 samples—Adapter set for NucleoVac 96 Vacuum Manifold available
- Can be used with standard manual and automated vacuum manifolds
- Optimized protocol for Cell-Free DNA BCT™ (Streck Tubes)
- The kit is fully automatable on many liquid handling robots
- The protocol follows state-of-the-art bind—wash—elute procedures
- Ready to use for downstream applications like real-time PCR or NGS
Specifications
Midi Spin Column | |
Blood Plasma | |
Cell-free DNA | |
DNA isolation from blood plasma | |
Fragment size: ≥ 50 bp | |
90 min/24 preps |
Silica Membrane Technology | |
Elution volume: 200 μL (140 μL final eluate) | |
Vacuum (e.g. NucleoVac 96 Vacuum Manifold and vacuum pump) | |
48 | |
1 - 5 mL | |
Depends on sample, typically: 0.1 - 100 ng DNA/mL plasma |