missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ DNA Insect, Mini kit for DNA from insects

Product Code. 15523626
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 15523626

Brand: Macherey-Nagel™ 740470.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Combines enzymatic lysis and mechanical disruption of cell walls for efficient isolation of genomic DNA from a wide variety of insects and crustacean samples (fresh, frozen, dried or ethanol preserved). Complete kit includes NucleoSpin™ Bead Tubes.

Isolation of genomic DNA from insects and crustaceans

  • Fast and convenient procedure combining enzymatic lysis and bead disruption
  • Suitable for any insect or crustacean (e.g., fruit fly, cricket, mealworm, midge larvae, mosquito)
  • High quality DNA from fresh, frozen, dried or ethanol preserved specimen samples
  • Bead Tubes included for difficult-to-lyse insects (due to exoskeletons and reinforced cell walls)
  • Efficient lysis – Compatible with the most common disruption devices
  • Typical downstream applications: multiplex PCR, Southern blotting, enzymatic reactions

Specifications

Content And Storage Room temperature (18 - 25 °C)
Format Mini Spin Column
Isolation Technology Silica MembraneTechnology and Mechanical (Bead Tube) Disruption
Sample Type Insect or Crustacean Specimens (Fresh, Frozen, Dried or Ethanol Preserved)
Efficiency Binding capacity: 60 μg
Elution Volume Elution volume: 25 - 200 μL
For Use With (Equipment) Disruption device (e.g. Vortex-Genie 2 with MN Bead Tube Holder), Centrifuge, Vortex mixer
For Use With (Application) Isolation of genomic DNA from insects and crustaceans
Includes NucleoSpin™ DNA Insect Columns, NucleoSpin™ Bead Tubes Type D, Collection Tubes (2 mL), Buffers
No. of Reactions 10
Purity A260/A280: 1.7 - 1.9
Sample Volume <40 mg fresh, frozen, dried or ethanol preserved insect or crustacean samples
Size Fragment size: 200 bp - approx. 50 kbp
Test Time 35 min/6 prep
Yield <25 μg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.