missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ DNA FFPE XS, Micro kit for DNA from FFPE
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
50 reactions
Description
Collection tubes (2mL) paraffin desolver, buffers, Proteinase K 250 preps Macherey Nagel
Specifications
Specifications
| Buffer | Yes |
| Final Product Type | NucleoSpin DNA FFPE XS |
| No. of Reactions | 50 |
| Product Type | NucleoSpin DNA FFPE XS |
| Sample Volume | ≤7 sections (10 μm) of 250 mm2 total area, <15 mg paraffin |
Safety and Handling
Product Identifier
- NucleoSpin DNA FFPE XS (50)
- Danger
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : paraffin disolver, proteinase k
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction