missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey Nagel Bioanalysis™ NucleoSpin Blood column
Specifications
Specifications
| Content And Storage | 18°C to 25°C |
| Format | Mini Prep |
| Sample Type | Blood, Body Fluids, Buffy Coat, Cells, Plasma, Platelets, Serum |
| For Use With (Application) | Isolation of DNA |
| Quantity | 250 Pk. |
| Sample Volume | 5 to 200 L whole blood (human or animal, fresh or frozen, treated with citrate, EDTA, heparin, or CPDA), buffy coat, platelets, body fluids, serum, plasma, < 5 x 106 human/animal cells cultured cells |
| Size | 200 bp–approx. 50 kbp |
| Target | DNA |
Safety and Handling
Product Identifier
- NucleoSpin Blood
- MIXTURE LIST-Contain: guanidine hydrochloride / 2-propanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?