Learn More
Macherey-Nagel™ NucleoSpin™ 96 Virus, 96-well kit for viral RNA/DNA purification
Description
Designed for the simultaneous purification of viral RNA and DNA. The kit combines the selectivity of well established silica membrane binding of nucleic acids with high throughput 96-well format. With the NucleoSpin™ 96 Virus method, RNA viruses are quickly and efficiently lysed by Lysis Buffer RAV1 which is a highly concentrated GITC solution. Compared to RNA viruses, DNA viruses (e.g., HBV) are usually more difficult to isolate and require a digestion of samples with Proteinase K which is provided in the kit. Lysis buffer and ethanol create appropriate conditions for binding of nucleic acids to the silica membrane of the NucleoSpin™ Virus Binding Plate. Carrier RNA included in Lysis Buffer RAV1 improves binding and recovery of low concentrated viral RNA/DNA. Contaminations (potential PCR inhibitors) like salts, metabolites, and soluble macromolecular cellular components are removed in washing steps with ethanolic Wash Buffer RAW and Buffer RAV3. The purified viral nucleic acids can be eluted in low salt buffer or water and are ready-to-use in subsequent downstream applications like RT-PCR or PCR.
- Silica membrane technology
- Sample material: Cell-free biological fluid, Plasma, Serum
- Fragment size: 100 bp to 50 kbp
- Theoretical binding capacity: 40 μL
- Elution volume: 70 to 100 μL
- Preparation time: 60 min./plate
Specifications
Specifications
| Content And Storage | 18°C to 25°C |
| Format | 96-well Plate |
| For Use With (Application) | Isolation of viral nucleic acids |
| No. of Reactions | 384 Preps |
| Sample Volume | <150 μL |
| Target | Viral RNA and DNA |
Safety and Handling
- Flammable liquid Category 3
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Your input is important to us. Please complete this form to provide feedback related to the content on this product.