missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Tissue, 96-well kit for DNA from cells and tissue

Product Code. 12788552
Change view
Click to view available options
No. of Reactions:
192 Preps
384 Preps
Unit Size:
192 reactions
384 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12788552 384 Preps 384 reactions
12768552 192 Preps 192 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12788552 Supplier Macherey-Nagel™ Supplier No. 740741.4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allround kit for purification of DNA from a broad range of samples in proven 96‐well plate format.

NucleoSpin™ 96 Tissue is designed for the rapid simultaneous preparation of highly pure genomic DNA from animal and human tissue, eg. mouse and rat tails.

From a 1.2 cm mouse tail tip section (age of mice 4 to 6 weeks), up to 30 to 40 μg of pure genomic DNA can be prepared (typical yields 15 to 25 μg). The obtained DNA can be used directly for PCR, Southern blotting or any kind of enzymatic reaction. NucleoSpin™ Tissue binding plates can be used under vacuum or centrifugation in a centrifuge with deep buckets allowing processing of NucleoSpin™ 96 well plates stacked on 96 well blocks. Up to 192 samples (two plates) can be processed in less than one hour (after lysis). With the NucleoSpin™ 96 Tissue method, lysis is achieved by incubation of the samples in a solution containing detergents and proteinase K at 56°C. Appropriate conditions for binding of DNA to the silica membranes of the NucleoSpin™ Tissue binding plate are created by addition of large amounts of chaotropic ions plus ethanol to the lysate. The binding process is reversible and specific to nucleic acids. Contaminants are removed by washing with two different buffers. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer.

Kit components: NucleoSpin™ Tissue binding plates, buffers, proteinase K, wash plates, square-well blocks, round-well blocks, racks with tube strips, cap strips, self-adhesive foil.

  • Silica membrane technology
  • Format: 96 well plates
  • Preparation time: 1 hour (after lysis)
  • Sample material: ≤20 mg tissue, 106 cultured or bacteria
  • Yield: 15 to 25 μg
  • Elution volume: 100 to 200 μL
  • Fragment size: 300 bp to 30 kb
  • No use of organic solvents

Specifications

Content And Storage 18°C to 25°C
Format 96-well Plate
For Use With (Application) Isolation of DNA
No. of Reactions 384 Preps
Sample Volume <20 mg human/animal tissue, <106 cells
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: guanidine hydrochloride, 2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.