missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 96 RNA Trace, 96‐well kit for RNA from cells and tissue
Click to view available options
:
4 x 96 Preps
Description
- Time-saving parallel isolation of total RNA
- rDNase included for efficient removal of genomic DNA
- Processing under vacuum or by centrifugation
- Suitable for manual and automated processing
- Innovative MN Wash Plate minimizes risk of cross-contamination
- RNA ready to use for any kind of enzymatic reaction
Specifications
Specifications
| Content And Storage | RNase-free rDNase at 4 °C, Other components at room temperature (18 - 25 °C) |
| Format | 96-well Plate |
| Isolation Technology | Silica Membrane |
| Sample Type | Cells, Tissue |
| Final Product Type | Total RNA |
| For Use With (Equipment) | Vacuum or Centrifuge |
| For Use With (Application) | Isolation of RNA from small sample amounts |
| Includes | NucleoSpin RNA Trace Binding Plates, MN Wash Plates, Square-well Blocks, Elution Plates U-bottom, buffers, RNase-free rDNase |
| No. of Samples | 4 x 96 |
| Sample Size | <2 x 106 cultured cells, <20 mg human/animal tissue |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction