Learn More
Macherey-Nagel™ NucleoSpin™ 96 RNA Blood
Complete kit with proven 96-well format for high throughput isolation of total RNA from fresh or frozen whole blood (human or animal) via direct total blood lysis that requires no selective erythrocyte lysis.
Marke: Macherey-Nagel™ 740225.2
Weitere Details : Weight : 0.65000kg
Beschreibung
NucleoSpin™ 96 RNA Blood Midi kit achieves superior RNA yield and quality in a format that provides convenient handling and flexible sample volume and throughput.
- Direct total blood lysis – very simple and convenient procedure
- No selective erythrocyte lysis at 4 °C – complete processing at RT
- Compatible with common blood collection tubes and anticoagulants (e.g., EDTA, citrate, and heparin)
- Suitable for fresh and frozen blood samples
- rDNase included for efficient removal of genomic DNA
- Processing under vacuum or by centrifugation
- Suitable for manual and automated (vacuum or centrifugation) processing
- Innovative MN Wash Plate minimizes risk of cross-contamination
Spezifikation
96-well Plate | |
Whole Blood (Fresh or Frozen) | |
Elution volume: 5 - 130 μL | |
Centrifuge or Vacuum | |
NucleoSpin RNA Blood Binding Plates, MN Wash Plates, Square-well Blocks, Round-well Blocks Low, Elution Plates U-bottom, Self-adhering Foil, buffers, RNase-free rDNase, Liquid Proteinase K | |
A260/A280: 1.9 - 2.1 | |
100 min/plate |
Silica Membrane Technology | |
Binding capacity: 100 μg | |
Total RNA | |
RNA isolation | |
2 x 96 | |
< 400 μL | |
1 - 8 μL |