missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Plasmid Core Kit, 96-well kit for plasmid DNA purification

Product Code. 15839107
Change view
Click to view available options
24 x 96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15839107 24 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15839107 Supplier Macherey-Nagel™ Supplier No. 740616.24
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rapid Mini preparation of plasmid DNA in proven 96‐well format.

NucleoSpin™ 96 Plasmid Core Kit for Isolation of Sequencing-Grade Plasmid DNA

  • Time-saving parallel isolation of plasmid DNA
  • Processing of up to 5 mL bacterial culture
  • NucleoSpin™ 96 Filter Plate for convenient filtration of bacterial lysates
  • Processing under vacuum or by centrifugation
  • Suitable for manual or automated processing under vacuum
  • Innovative MN Wash Plate minimizes risk of cross-contamination (not included in Core Kit, available separately or in full kit)

Specifications

Content And Storage Room temperature (18 - 25 °C)
Format 96-well Plate
Isolation Technology Silica Membrane Technology
Sample Type E. coli Culture
Efficiency Binding capacity: 20 μg
Elution Volume Elution volume: 75 - 150 μL
Final Product Type Sequencing-grade plasmid DNA
For Use With (Equipment) Liquid handling system, Vacuum
For Use With (Application) High throughput DNA isolation
No. of Samples 24 x 96
Sample Volume 1 - 5 mL
Size < 15 kbp
Test Time 45 min/plate
Yield 4 - 6 μg/mL culture
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin 96 Plasmid Core Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash with water thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide, dodecyl sulphate sodium salt, guanidine hydrochloride, Rnase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.