missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Plant II, 96-well kit for DNA from plants

Product Code. 12783560
Change view
Click to view available options
Quantity:
192 preps
2304 preps
384 preps
Unit Size:
192 reactions
2304 reactions
384 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12783560 384 preps 384 reactions
12763560 192 preps 192 reactions
12773560 2304 preps 2304 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12783560 Supplier Macherey-Nagel™ Supplier No. 740663.4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapid isolation of DNA from a multitude of plant samples in proven 96‐well plate format.

NucleoSpin™ 8/96 Plant is designed for the isolation of genomic DNA from plant tissue (e.g., Arabidopsis, tobacco or maize).

Depending on the individual sample (20mg to 100 mg) typical yields are in the range from 5 to 30μg DNA. For RNA-rich samples, it is recommended to add RNase to the lysis buffer or perform a subsequent RNase treatment. The general isolation procedure ensures lysis of plant material with a CTAB lysis buffer developed especially for plant samples. CTAB is removed completely in the subsequent washing steps and thus does not interfere with downstream PCR*.The binding process is reversible and specific for nucleic acids. Contaminations are removed by washing. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer. The obtained DNA can be used directly for PCR*, Southern blotting or any kind of enzymatic reaction.NucleoSpin™ 8/96 Plant can be used in a centrifuge with deep buckets allowing processing of NucleoSpin™ 8 well strips/96 well plates stacked on 96 well blocks. NucleoSpin™ 8/96 can also be used under vacuum, manually in the NucleoVac 96, or automated on many common liquid handling instruments. Up to 192 samples (two full 96 well plates) can be processed in 2 to 3hrs depending on equipment used for pipetting and homogenisation.

Required hardware

Centrifugation

For centrifuge processing of NucleoSpin 8/96 plant kits a microtiterplate centrifuge is required which is able to accommodate the NucleoSpin Plant strips/plate stacked on a round or square well block or rack with tube strips and reaches accelerations of 5,600xg to 6,000xg (bucket height: 85mm). For further information on suitable centrifuges please contact our Technical Service team. For centrifuge processing of the 8 well Strips, the reusable Starter Set C, containing column holders C, dummy strips, MN square well blocks, and tube strips, is required.

Vacuum processing

The NucleoSpin 8/96 plant kits can be used manually with the NucleoVac 96 vacuum manifold. Additionally, a suitable centrifuge for sample preparation steps may be required.For vacuum processing of the 8 well strips the reusable Starter Set A, containing column holders A and dummy strips is required. For automation on laboratory platforms with standard 96 well plate vacuum chambers the use of the Starter Set A is also required.

Kit components

NucleoSpin™ 8 Plant - NucleoSpin™ plant binding strips, wash plates, round well blocks, racks with tube strips, cap strips, gas permeable foil, self-adhering foil, buffers, RNase A. NucleoSpin™ 96 Plant - NucleoSpin™ plant binding plates, square well blocks, racks with tube strips, cap strips, gas-permeable foil, buffers, RNase A.

  • Silica membrane technology
  • Two formats: 8 well strip or 96 well plates
  • Yield: 5μg to 30μg
  • Sample material: 20μg to 100μg plant tissue or cells
  • Elution volume: 100μL to 200μL
  • Binding capacity: 25μg (8 well format)/30μg (96 well format)
  • Preparation time: 120min for 12 strips/one 96 well plate
  • MN wash plate minimises cross-contamination

For isolation of genomic DNA from plant cells/tissue in a 8 well strip format or 96 well format.

Specifications

Format 96-well Plate
Isolation Technology Silica Membrane Technology
For Use With (Application) For Manual and Automated Isolation of Genomic DNA
No. of Reactions 4 x 96
Product Type NucleoSpin™ 96 Plant Binding Plate
Quantity 384 preps
Test Time 60min.
Yield 1 to 30μg
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.