missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 PCR Clean-up, 96-well kit for PCR clean up

Product Code. 12755214
Click to view available options
No. of Reactions:
192 Preps
2304 Preps
384 Preps
96 Preps
Unit Size:
192 reactions
2304 reactions
384 reactions
96 reactions
This item is not returnable. View return policy

Product Code. 12755214

Brand: Macherey-Nagel™ 740658.4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NucleoSpin™ 96 Gel and PCR Clean-Up Kit provides time saving medium to high throughput PCR clean up.

The NucleoSpin™ 96 PCR Clean-up kit allows direct clean-up of PCR reaction mixtures. Within the procedure the addition of chaotropic salt (Buffer NT) allows a reversible adsorption of the PCR products to the silica membrane of the NucleoSpin™ 96 PCR Clean-up Binding Plates. High purity of the PCR-products is achieved by complete removal of primers, primer-dimers, salts, nucleotides, and proteins (e.g., polymerases, BSA) in subsequent washing steps using Buffer NT3. Highly pure PCR products are finally eluted with Elution Buffer NE (5 mM Tris/HCl, pH 8.5) or water (pH 8.5), and can be used directly for further applications.

  • Silica membrane technology
  • Processing: manual/automated, vacuum/centrifugation
  • Sample material: < 10 0μL PCR reaction mixture
  • Elution volume: 75 to 150 μL
  • Binding capacity: 15 μg
  • Preparation time: 45 min./plate
  • Fragment size: 65 bp to 10 kb

Specifications

Content And Storage 18°C to 25°C
Format 96-well Plate
For Use With (Application) PCR clean up
No. of Reactions 384 Preps
Sample Volume <100 μL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.