missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Food, 96-well kit for DNA from food

Product Code. 12738632
Click to view available options
No. of Reactions:
24 x 96
2x 96
4 x 96
Packungsgröße:
192 reactions
2304 reactions
384 reactions
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 12738632

Marke: Macherey-Nagel™ 740976.24

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

NucleoSpin 96 Food, 96-well kit for DNA from food , MN

The NucleoSpin™ Food and NucleoSpin™ Food 8/96 systems guarantee good recovery rates even for small genomic DNA fragments (<1kb) out of processed, complex food matrices, which generally have very low DNA contents as well as poor quality, degraded DNA.

After the food samples have been homogenised, the DNA is extracted with lysis buffers containing chaotropic salts, denaturing agents and detergents. The standard isolation ensures lysis using buffer CF, which was especially developed in cooperation with GEN-IAL, Troisdorf, Germany (patent pending). Lysis mixtures should be cleared by centrifugation or filtration in order to remove contaminants and residual cellular debris. The clear supernatant is then mixed with the binding buffer and ethanol to create conditions for optimal binding to the NucleoSpin™ silica membrane, selected for this purpose due to its unique DNA binding properties. After washing with two different buffers for efficient of potential PCR* inhibitors, DNA can be eluted in low salt buffer or water. Resulting eluates are ready-to-use in all types of subsequent detection methods, especially real-time and basic PCR* technologies and can be used for the identification of GMO-DNA or animal components in food and feed.

Kit components

NucleoSpin™ Food - NucleoSpin™ Food columns, collecting tubes 2mL, buffers, proteinase K. NucleoSpin™ 8 Food - NucleoSpin™ Food binding strips, round-well blocks, MN wash plates, racks with tube strips, cap strips, gas-permeable foil, buffers, proteinase K.NucleoSpin™ 96 Food - NucleoSpin™ 96 Food binding plates, round-well blocks, MN square-well blocks, MN wash plates, racks with tube strips, gas-permeable foil, buffers, proteinase K.

  • Silica membrane technology
  • Available formats:
    • NucleoSpin - mini spin column
    • NucleoSpin 8 - 8 well strip
    • NucleoSpin 96 - 96 well plate
  • Yield: 0.1μg to 10μg
  • Sample material: ≤200μg food/feed
  • Elution volume: 100μL (mini spin column) 100μL to 200μL (8 or 96 well formats)
  • Preparation time: 30 min./6 preps (mini spin columns) <60 min./8 strips (excluding lysis) <120 min./96 well plate (excluding lysis)
  • Manual or automated processing, vacuum/centrifugation
  • No use of organic solvents

Designed for medium to high-throughput isolation of GMO-DNA or animal components in food and feed.

Spezifikation

Format 96-well Plate
Isolation Technology Silica Membrane Technology
For Use With (Application) For Manual and Automated Isolation of DNA
No. of Reactions 24 x 96
Product Type NucleoSpin™ 96 Food Binding Plate
Sample Volume <200 mg
Test Time 120min.
Yield 0.1 to 10 μg (200 mg food)
compliance-icons compliance-icons compliance-icons
Gefahrenkategorie
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt