missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Food, 96-well kit for DNA from food

Product Code. 12728632
Change view
Click to view available options
No. of Reactions:
24 x 96
2x 96
4 x 96
Unit Size:
192 reactions
2304 reactions
384 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12728632 2x 96 192 reactions
12738632 24 x 96 2304 reactions
12762573 4 x 96 384 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12728632 Supplier Macherey-Nagel™ Supplier No. 740976.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NucleoSpin 96 Food, 96-well kit for DNA from food , MN

The NucleoSpin™ Food and NucleoSpin™ Food 8/96 systems guarantee good recovery rates even for small genomic DNA fragments (<1kb) out of processed, complex food matrices, which generally have very low DNA contents as well as poor quality, degraded DNA.

After the food samples have been homogenised, the DNA is extracted with lysis buffers containing chaotropic salts, denaturing agents and detergents. The standard isolation ensures lysis using buffer CF, which was especially developed in cooperation with GEN-IAL, Troisdorf, Germany (patent pending). Lysis mixtures should be cleared by centrifugation or filtration in order to remove contaminants and residual cellular debris. The clear supernatant is then mixed with the binding buffer and ethanol to create conditions for optimal binding to the NucleoSpin™ silica membrane, selected for this purpose due to its unique DNA binding properties. After washing with two different buffers for efficient of potential PCR* inhibitors, DNA can be eluted in low salt buffer or water. Resulting eluates are ready-to-use in all types of subsequent detection methods, especially real-time and basic PCR* technologies and can be used for the identification of GMO-DNA or animal components in food and feed.

Kit components

NucleoSpin™ Food - NucleoSpin™ Food columns, collecting tubes 2mL, buffers, proteinase K. NucleoSpin™ 8 Food - NucleoSpin™ Food binding strips, round-well blocks, MN wash plates, racks with tube strips, cap strips, gas-permeable foil, buffers, proteinase K.NucleoSpin™ 96 Food - NucleoSpin™ 96 Food binding plates, round-well blocks, MN square-well blocks, MN wash plates, racks with tube strips, gas-permeable foil, buffers, proteinase K.

  • Silica membrane technology
  • Available formats:
    • NucleoSpin - mini spin column
    • NucleoSpin 8 - 8 well strip
    • NucleoSpin 96 - 96 well plate
  • Yield: 0.1μg to 10μg
  • Sample material: ≤200μg food/feed
  • Elution volume: 100μL (mini spin column) 100μL to 200μL (8 or 96 well formats)
  • Preparation time: 30 min./6 preps (mini spin columns) <60 min./8 strips (excluding lysis) <120 min./96 well plate (excluding lysis)
  • Manual or automated processing, vacuum/centrifugation
  • No use of organic solvents

Designed for medium to high-throughput isolation of GMO-DNA or animal components in food and feed.

Specifications

Format 96-well Plate
Isolation Technology Silica Membrane Technology
For Use With (Application) For Manual and Automated Isolation of DNA
No. of Reactions 2x 96
Product Type NucleoSpin™ 96 Food Binding Plate
Sample Volume <200 mg
Test Time 120min.
Yield 0.1 to 10 μg (200 mg food)
compliance-icons compliance-icons compliance-icons
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.