missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 cfDNA, 96-well kit for cell-free DNA from plasma

Product Code. 15698455
Click to view available options
Quantity:
384 preps
96 preps
Product Type:
Core Kit
Full Kit
Unit Size:
1 set
4 sets
This item is not returnable. View return policy

Product Code. 15698455

Brand: Macherey-Nagel™ 740874.4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High throughput solution for isolation of cell-free circulating DNA from up to 2 mL blood plasma in 96-well format. Available in full or core kits, with one or four plates.

NucleoSpin™ 96 DNA Plasma Kits

  • High throughput solution for cell-free DNA isolation
  • Fragmented DNA of 50 bp and larger can be purified with high efficiency
  • Manual or automated processing
  • Protocol follows state-of-the-art bind-wash-elute procedures
  • Optimized for Cell-Free DNA BCT™ ('Streck Tubes'), or for standard EDTA blood draw tubes
  • Core Kits include basic content focused on automation platforms
  • Typical downstream applications: NGS, qPCR, ddPCR

Specifications

Content And Storage Room temperature (18-25 °C)
Format 96-well Plate
Isolation Technology Silica Membrane Technology
Sample Type Blood Plasma (esp. human)
Elution Volume Elution volume: 100 μL
For Use With (Equipment) Vacuum manifold/pump
For Use With (Application) Isolation of cell-free, circulating DNA from plasma
Includes NucleoSpin Plasma Binding Plate, buffers, Proteinase K
No. of Samples 4 x 96
Product Type Core Kit
Quantity 384 preps
Sample Volume 0.5 to 2 mL
Test Time 90 min/96 preps
Yield Depends on sample source, storage, and quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.