missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 96 cfDNA, 96-well kit for cell-free DNA from plasma
Click to view available options
Quantity:
384 preps
96 preps
Product Type:
Core Kit
Full Kit
Unit Size:
1 set
4 sets
Description
NucleoSpin™ 96 DNA Plasma Kits
- High throughput solution for cell-free DNA isolation
- Fragmented DNA of 50 bp and larger can be purified with high efficiency
- Manual or automated processing
- Protocol follows state-of-the-art bind-wash-elute procedures
- Optimized for Cell-Free DNA BCT™ ('Streck Tubes'), or for standard EDTA blood draw tubes
- Core Kits include basic content focused on automation platforms
- Typical downstream applications: NGS, qPCR, ddPCR
Specifications
Specifications
| Content And Storage | Room temperature (18-25 °C) |
| Format | 96-well Plate |
| Isolation Technology | Silica Membrane Technology |
| Sample Type | Blood Plasma (esp. human) |
| Elution Volume | Elution volume: 100 μL |
| For Use With (Equipment) | Vacuum manifold/pump |
| For Use With (Application) | Isolation of cell-free, circulating DNA from plasma |
| Includes | NucleoSpin Plasma Binding Plate, buffers, Proteinase K |
| No. of Samples | 1 x 96 |
| Product Type | Core Kit |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction