missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Blood QuickPure, 96-well kit for quick DNA extraction from blood

Product Code. 12758522
Change view
Click to view available options
No. of Reactions:
192 Preps
2304 Preps
384 Preps
Unit Size:
192 reactions
2304 reactions
384 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12758522 192 Preps 192 reactions
12778522 384 Preps 384 reactions
12768522 2304 Preps 2304 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12758522 Supplier Macherey-Nagel™ Supplier No. 740667.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ultra-fast isolation of highly concentrated DNA from blood in proven 96‐well plate format.

NucleoSpin™ Blood QuickPure binding plates are to be used in a centrifuge with deep buckets allowing processing of NucleoSpin™ 96 well plates stacked on 96 well blocks. The method is based on a specially treated silica membrane which speeds up the procedure by a combined washing/drying step. The number of washing and drying steps is reduced from 3 to 1. Therefore the hands-on time is minimised. Lysis is achieved by incubation of whole blood in a solution containing large amounts of chaotropic ions in the presence of proteinase K. Appropriate conditions for binding of DNA to the silica membrane of the NucleoSpin™ Blood QuickPure binding strips are created by addition of ethanol to the lysate. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer and can be used directly for PCR, Southern blotting or any kind of enzymatic reaction. The kit allows isolation of 4 μg to 6 μg of pure genomic DNA from 200 μL whole blood with an A260/280 ratio between 1.60 and 1.90 and a typical concentration of 40 to 60 ng/μL.Kit components: NucleoSpin™ Blood QuickPure binding plates, buffers, proteinase K, accessories.

  • Silica membrane technology
  • Manual processing
  • Yield: 4 to 6 μg
  • Fragment size: 300 bp to 30 kb
  • Sample material: human blood or other body fluid (200 μL/300 μL), 107 lymphocytes
  • Elution volume: 75 to 100 μL
  • Binding capacity: 60 μg
  • Preparation time: 30 min./prep

Specifications

Content And Storage 18°C to 25°C
Format 96-well Plate
For Use With (Application) Isolation of DNA
No. of Reactions 192 Preps
Target DNA
compliance-icons compliance-icons
Hazard Category
  • Skin sensitiser Category 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.