Learn More
Macherey-Nagel™ NucleoSpin™ 96 Blood QuickPure, 96-well kit for quick DNA extraction from blood
Description
NucleoSpin™ Blood QuickPure binding plates are to be used in a centrifuge with deep buckets allowing processing of NucleoSpin™ 96 well plates stacked on 96 well blocks. The method is based on a specially treated silica membrane which speeds up the procedure by a combined washing/drying step. The number of washing and drying steps is reduced from 3 to 1. Therefore the hands-on time is minimised. Lysis is achieved by incubation of whole blood in a solution containing large amounts of chaotropic ions in the presence of proteinase K. Appropriate conditions for binding of DNA to the silica membrane of the NucleoSpin™ Blood QuickPure binding strips are created by addition of ethanol to the lysate. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer and can be used directly for PCR, Southern blotting or any kind of enzymatic reaction. The kit allows isolation of 4 μg to 6 μg of pure genomic DNA from 200 μL whole blood with an A260/280 ratio between 1.60 and 1.90 and a typical concentration of 40 to 60 ng/μL.Kit components: NucleoSpin™ Blood QuickPure binding plates, buffers, proteinase K, accessories.
- Silica membrane technology
- Manual processing
- Yield: 4 to 6 μg
- Fragment size: 300 bp to 30 kb
- Sample material: human blood or other body fluid (200 μL/300 μL), 107 lymphocytes
- Elution volume: 75 to 100 μL
- Binding capacity: 60 μg
- Preparation time: 30 min./prep
Specifications
Specifications
| Content And Storage | 18°C to 25°C |
| Format | 96-well Plate |
| For Use With (Application) | Isolation of DNA |
| No. of Reactions | 384 Preps |
| Target | DNA |
Safety and Handling
- Skin sensitiser Category 1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.