missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Blood, 96-well kit for DNA from blood

Product Code. 12766523
Click to view available options
No. of Reactions:
1 x 96
24 x 96
Unit Size:
2304 reactions
96 reactions
This item is not returnable. View return policy

Product Code. 12766523

Brand: Macherey-Nagel™ 740665.24

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Versatile purification of high quality DNA from whole blood in proven 96‐well plate format.

The NucleoSpin 8 Extract II kits allow the rapid clean-up of up to 15μg of DNA per preparation with high DNA recovery and reproducible yields. After purification with NucleoSpin 8 Extract II the highly pure PCR products are ready for immediate use in many applications, such as radioactive and fluorescent dye sequencing, including capillary sequencing, restriction analysis, ligation, and microarray analysis. NucleoSpin Extract II Binding Strips/Plates can be used under vacuum or by centrifugation. The NucleoSpin 8 Extract II kit uses advanced silica-membrane technology. The addition of a binding buffer containing chaotropic ions leads to a reversible adsorption of DNA to the NucleoSpin 8 Extract II silica membrane. PCR products bind to the silica membrane under optimised conditions, while contaminants like primers, salts, nucleotides, and proteins are completely removed by effective washing. Highly pure PCR products are eluted with low-salt elution buffer (5mM Tris/HCl pH8.5) or water (pH8.0 to 8.5) and are ready-to-use directly for subsequent applications.

Required hardware

Centrifugation

For centrifuge processing of NucleoSpin 8 Extract II kits a microtiterplate centrifuge is required which is able to accommodate the NucleoSpin Extract II Strips/Plate stacked on a round- or square-well block or rack with tube strips and reaches accelerations of 5,600 - 6,000 x g (bucket height: 85mm).For centrifuge processing of the 8-well strips the reusable starter set C, containing column holders C, dummy strips, MN square-well blocks, tube strips is required.

Vacuum processing

The NucleoSpin 8 Extract II kits can be used manually with the NucleoVac 96 vacuum manifold.For vacuum processing of the 8-well strips the reusable starter set A, containing column holders A and dummy strips is required.Kit components: NucleoSpin Extract binding strips, MN wash plate, racks with tube strips, cap strips, buffers.

  • Silica membrane technology
  • Format: 8-well strips
  • Yield: 75% to 95%
  • Sample material: <100μL PCR reaction mixture
  • Fragment size: 65bp to 10kb
  • Elution volume:75μL to 150μL
  • Binding capacity: 15μg
  • Preparation time: 30min./6 strips
  • Format: mini spin column

For clean-up of PCR* products.

Specifications

Format 96-well Plate
Isolation Technology Silica Membrane Technology
For Use With (Application) For Manual and Automated Isolation of Genomic DNA
No. of Reactions 24 x 96
Product Type NucleoSpin™ 96 Blood Binding Plate
Test Time 70min.
Yield 4 to 6μg
compliance-icons compliance-icons compliance-icons
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.