Learn More
Macherey-Nagel™ NucleoSpin™ 96 Blood, 96-well kit for DNA from blood
Description
The NucleoSpin 8 Extract II kits allow the rapid clean-up of up to 15μg of DNA per preparation with high DNA recovery and reproducible yields. After purification with NucleoSpin 8 Extract II the highly pure PCR products are ready for immediate use in many applications, such as radioactive and fluorescent dye sequencing, including capillary sequencing, restriction analysis, ligation, and microarray analysis. NucleoSpin Extract II Binding Strips/Plates can be used under vacuum or by centrifugation. The NucleoSpin 8 Extract II kit uses advanced silica-membrane technology. The addition of a binding buffer containing chaotropic ions leads to a reversible adsorption of DNA to the NucleoSpin 8 Extract II silica membrane. PCR products bind to the silica membrane under optimised conditions, while contaminants like primers, salts, nucleotides, and proteins are completely removed by effective washing. Highly pure PCR products are eluted with low-salt elution buffer (5mM Tris/HCl pH8.5) or water (pH8.0 to 8.5) and are ready-to-use directly for subsequent applications.
Required hardware
Centrifugation
For centrifuge processing of NucleoSpin 8 Extract II kits a microtiterplate centrifuge is required which is able to accommodate the NucleoSpin Extract II Strips/Plate stacked on a round- or square-well block or rack with tube strips and reaches accelerations of 5,600 - 6,000 x g (bucket height: 85mm).For centrifuge processing of the 8-well strips the reusable starter set C, containing column holders C, dummy strips, MN square-well blocks, tube strips is required.
Vacuum processing
The NucleoSpin 8 Extract II kits can be used manually with the NucleoVac 96 vacuum manifold.For vacuum processing of the 8-well strips the reusable starter set A, containing column holders A and dummy strips is required.Kit components: NucleoSpin Extract binding strips, MN wash plate, racks with tube strips, cap strips, buffers.
- Silica membrane technology
- Format: 8-well strips
- Yield: 75% to 95%
- Sample material: <100μL PCR reaction mixture
- Fragment size: 65bp to 10kb
- Elution volume:75μL to 150μL
- Binding capacity: 15μg
- Preparation time: 30min./6 strips
- Format: mini spin column
For clean-up of PCR* products.
Specifications
Specifications
| Format | 96-well Plate |
| Isolation Technology | Silica Membrane Technology |
| For Use With (Application) | For Manual and Automated Isolation of Genomic DNA |
| No. of Reactions | 1 x 96 |
| Product Type | NucleoSpin™ 96 Blood Binding Plate |
| Test Time | 70min. |
| Yield | 4 to 6μg |
Safety and Handling
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Your input is important to us. Please complete this form to provide feedback related to the content on this product.