Learn More
Macherey-Nagel™ NucleoSpin™ 8 PCR Clean-up, 8-well kit for PCR clean up
Description
The NucleoSpin 8 Extract II kits allow the rapid clean-up of up to 15?g of DNA per preparation with high DNA recovery and reproducible yields. After purification with NucleoSpin 8 Extract II the highly pure PCR products are ready for immediate use in many applications, such as radioactive and fluorescent dye sequencing, including capillary sequencing, restriction analysis, ligation, and microarray analysis. NucleoSpin Extract II Binding Strips/Plates can be used under vacuum or by centrifugation. The NucleoSpin 8 Extract II kit uses advanced silica-membrane technology. The addition of a binding buffer containing chaotropic ions leads to a reversible adsorption of DNA to the NucleoSpin 8 Extract II silica membrane. PCR products bind to the silica membrane under optimised conditions, while contaminants like primers, salts, nucleotides, and proteins are completely removed by effective washing. Highly pure PCR products are eluted with low-salt elution buffer (5mM Tris/HCl pH8.5) or water (pH8.0 to 8.5) and are ready-to-use directly for subsequent applications.
Required hardware
Centrifugation
For centrifuge processing of NucleoSpin 8 Extract II kits a microtiterplate centrifuge is required which is able to accommodate the NucleoSpin Extract II Strips/Plate stacked on a round- or square-well block or rack with tube strips and reaches accelerations of 5,600 - 6,000 x g (bucket height: 85mm).For centrifuge processing of the 8-well strips the reusable starter set C, containing column holders C, dummy strips, MN square-well blocks, tube strips is required.
Vacuum processing
The NucleoSpin 8 Extract II kits can be used manually with the NucleoVac 96 vacuum manifold.For vacuum processing of the 8-well strips the reusable starter set A, containing column holders A and dummy strips is required.Kit components: NucleoSpin Extract binding strips, MN wash plate, racks with tube strips, cap strips, buffers.
- Silica membrane technology
- Format: 8-well strips
- Yield: 75% to 95%
- Sample material: <100μL PCR reaction mixture
- Fragment size: 65bp to 10kb
- Elution volume:75μL to 150μL
- Binding capacity: 15μg
- Preparation time: 30 min./6 strips
- Format: mini spin column
For clean-up of PCR* products.
Specifications
Specifications
| Format | 8-well Strip |
| Isolation Technology | Silica Membrane Technology |
| For Use With (Application) | For Manual and Automated Purification of PCR Products |
| No. of Reactions | 12 x 8 |
| Product Type | NucleoSpin™ 8 PCR Clean-up Kit |
| Sample Volume | <100 μL |
| Test Time | 30min. |
Safety and Handling
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- MIXTURE LIST-Contain: guanidinium thiocyanate
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.