missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 8 Virus Strips for viral RNA/DNA purification
Click to view available options
Quantity:
12 x 8-well Strips
60 x 8-Well Strips
Unit Size:
Pack of 12
Pack of 60
Description
NucleoSpin™ 8 Virus Kit—Isolate viral RNA and DNA from serum and plasma
- Parallel isolation using a flexible 8-well strip format for varying throughput
- For processing under vacuum or by centrifugation
- Suitable for manual and automated processing
- Innovative MN Wash Plate minimizes risk of cross-contamination
- Wide range of viral samples: HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1, H1N1
- Typical downstream applications: PCR, RT-PCR, or any kind of enzymatic reaction
Specifications
Specifications
| Format | 8-well Strip |
| Isolation Technology | Silica Membrane Technology |
| Sample Type | Cell-free Biological Fluids, Plasma, Serum |
| Efficiency | Binding capacity: 40 μg |
| Elution Volume | Elution volume: 70 - 100 μL |
| For Use With (Equipment) | NucleoSpin 8 Virus full or core kit |
| For Use With (Application) | Parallel isolation of viral RNA and DNA |
| No. of Samples | 8/Strip |
| Quantity | 60 x 8-Well Strips |
| Sample Volume | <150 μL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction