Learn More
Macherey-Nagel™ NucleoSpin™ 8 Soil
Flexible 8-well strip format for varying throughput in the isolation of total DNA from microorganisms in soil and sediment samples.
Brand: Macherey-Nagel™ 740779
Additional Details : Weight : 1.15000kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
NucleoSpin™ 8 Soil
- High quality DNA from diverse soil samples
- Different lysis buffer options for optimal processing of various soil sample composition
- Ceramic beads in bead tubes for most efficient lysis and no cross-contamination during lysis
- NucleoSpin™ Inhibitor Removal Strips/Plate for eliminating all PCR inhibitors, such as humic substances
- Typical downstream applications: qPCR, PCR, micro array, any kind of enzymatic reaction
Specifications
8-well Strip | |
Sediment, Sludge, Soil | |
Elution volume: 100 - 200 μL | |
Vacuum or centrifuge | |
NucleoSpin Soil Binding Strips, NucleoSpin Bead Tubes, NucleoSpin Inhibitor Removal Strips, MN Wash Plates, MN Square-well Blocks, Rack of Tube Strips, buffers | |
8/Strip | |
< 500 mg | |
2 - 10 μg |
Silica Membrane Technology | |
Binding capacity: 50 μg | |
Genomic DNA | |
Isolation of DNA from microorganisms in soil and sediment | |
12 x 8 Preps | |
A260/A280: 1.6 - 1.8 | |
150 min/6 strips |
Safety and Handling
- NucleoSpin 8 Soil
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash with water thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate; guanidine hydrochloride, 2-propanol