missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 8 PCR Clean-up, 8-well kit for PCR clean up

Product Code. 12798522
Change view
Click to view available options
:
8 well binding strips, MN wash plate, rack of tube strips, buffers (formerly known as NucleoSpin™ 8 Extract II) Macherey-Nagel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12798522 8 well binding strips, MN wash plate, rack of tube strips, buffers (formerly known as NucleoSpin™ 8 Extract II) Macherey-Nagel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12798522 Supplier Macherey-Nagel™ Supplier No. 740668.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Time saving medium to high throughput PCR clean up.

The NucleoSpin 8 Extract II kits allow the rapid clean-up of up to 15μg of DNA per preparation with high DNA recovery and reproducible yields. After purification with NucleoSpin 8 Extract II the highly pure PCR products are ready for immediate use in many applications, such as radioactive and fluorescent dye sequencing, including capillary sequencing, restriction analysis, ligation, and microarray analysis. NucleoSpin Extract II Binding Strips/Plates can be used under vacuum or by centrifugation. The NucleoSpin 8 Extract II kit uses advanced silica-membrane technology. The addition of a binding buffer containing chaotropic ions leads to a reversible adsorption of DNA to the NucleoSpin 8 Extract II silica membrane. PCR products bind to the silica membrane under optimised conditions, while contaminants like primers, salts, nucleotides, and proteins are completely removed by effective washing. Highly pure PCR products are eluted with low-salt elution buffer (5mM Tris/HCl pH8.5) or water (pH8.0 to 8.5) and are ready-to-use directly for subsequent applications.

Required hardware

Centrifugation

For centrifuge processing of NucleoSpin 8 Extract II kits a microtiterplate centrifuge is required which is able to accommodate the NucleoSpin Extract II Strips/Plate stacked on a round- or square-well block or rack with tube strips and reaches accelerations of 5,600 - 6,000 x g (bucket height: 85mm).For centrifuge processing of the 8-well strips the reusable starter set C, containing column holders C, dummy strips, MN square-well blocks, tube strips is required.

Vacuum processing

The NucleoSpin 8 Extract II kits can be used manually with the NucleoVac 96 vacuum manifold.For vacuum processing of the 8-well strips the reusable starter set A, containing column holders A and dummy strips is required.Kit components: NucleoSpin Extract binding strips, MN wash plate, racks with tube strips, cap strips, buffers.

  • Silica membrane technology
  • Format: 8-well strips
  • Yield: 75% to 95%
  • Sample material: <100μL PCR reaction mixture
  • Fragment size: 65bp to 10kb
  • Elution volume:75μL to 150μL
  • Binding capacity: 15μg
  • Preparation time: 30 min./6 strips
  • Format: mini spin column

For clean-up of PCR* products.

Specifications

Format 8-well Strip
Isolation Technology Silica Membrane Technology
For Use With (Application) For Manual and Automated Purification of PCR Products
No. of Reactions 60 x 8
Product Type NucleoSpin™ 8 PCR Clean-up Binding Strip
Sample Volume <100 μL
Test Time 30min.
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: guanidinium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.