missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 8 Plasmid, 8-well kit for plasmid DNA

Product Code. 12766513
Change view
Click to view available options
No. of Reactions:
12 x 8 Preps
60 x 8 Preps
Unit Size:
480 reactions
96 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12766513 60 x 8 Preps 480 reactions
12756513 12 x 8 Preps 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12766513 Supplier Macherey-Nagel™ Supplier No. 740621.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for manual or automated purification of high-copy plasmid DNA from E.coli. Macherey-Nagel™ NucleoSpin™ 8 Pasmid Kit is an easy-to-use kit that supplies all core and accessory strips, buffers and RNase A for maximum convenience. The 8-well strip format is ideal for varying throughput.

NucleoSpin™ 8 Pasmid kits enable variable-throughput isolation of sequencing-grade plasmid DNA from E. coli. The flexible 8-well strip format allows for automation on common liquid handling instruments or for manual processing without time-consuming centrifugation.

Up to 8 samples per strip can be parallel-processed simultaneously, typically within less than 45 minutes. Uses include isolation of high-copy plasmids, purification of cosmids and reliable downstream applications, such as cloning, sequencing, PCR, transformation, and restriction analysis.

  • Time-saving parallel isolation of plasmid DNA using silica-membrane technology
  • Processing of up to 5 mL bacterial culture, with a yield up to 6 μg/mL
  • 8-well filter strip for convenient filtration of bacterial lysates
  • Suitable for manual or automated processing under vacuum
  • Innovative MN Wash Plate minimizes risk of cross-contamination

Specifications

Format 8-well Strip
Sample Type E. coli Culture
Efficiency Binding capacity: 20μg
Elution Volume Elution volume: 75–150μL
Final Product Type Sequencing grade plasmid DNA
For Use With (Equipment) Liquid handling instruments
For Use With (Application) Manual and automated processing under vacuum
Includes NucleoSpin Plasmid Filter Strips (60), NucleoSpin Plasmid Binding Strips (60), MN Wash Plate (5), Culture plate with Gas-permeable Foil (5), Rack of Tube Strips (5), RNase A (60mg), Resuspension Buffer A1 (150mL), Lysis Buffer A2 (150mL), Neutralization Buffer A3 (200mL), Wash Buffer AW (500mL), Wash Buffer A4 Concentrate (4 x 100mL), Elution Buffer AE (125mL)
No. of Reactions 60 x 8 Preps
Product Type NucleoSpin™ 8 Plasmid Kit
Quantity 1 Kit
Purity A260/A280: 1.70–1.85
Sample Volume 1–5mL
Size <15 kbp
Test Time Preparation time: 45 min. / 6 strips
Yield 4–6μg plasmid per 1mL E. coli culture
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt; guanidine hydrochloride; 2-
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.