missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 8 Plasmid, 8-well kit for plasmid DNA

Product Code. 12756513
Click to view available options
No. of Reactions:
12 x 8 Preps
60 x 8 Preps
Unit Size:
480 reactions
96 reactions
This item is not returnable. View return policy

Product Code. 12756513

Brand: Macherey-Nagel™ 740621

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for manual or automated purification of high-copy plasmid DNA from E.coli. Macherey-Nagel™ NucleoSpin™ 8 Pasmid Kit is an easy-to-use kit that supplies all core and accessory strips, buffers and RNase A for maximum convenience. The 8-well strip format is ideal for varying throughput.

NucleoSpin™ 8 Pasmid kits enable variable-throughput isolation of sequencing-grade plasmid DNA from E. coli. The flexible 8-well strip format allows for automation on common liquid handling instruments or for manual processing without time-consuming centrifugation.

Up to 8 samples per strip can be parallel-processed simultaneously, typically within less than 45 minutes. Uses include isolation of high-copy plasmids, purification of cosmids and reliable downstream applications, such as cloning, sequencing, PCR, transformation, and restriction analysis.

  • Time-saving parallel isolation of plasmid DNA using silica-membrane technology
  • Processing of up to 5 mL bacterial culture, with a yield up to 6 μg/mL
  • 8-well filter strip for convenient filtration of bacterial lysates
  • Suitable for manual or automated processing under vacuum
  • Innovative MN Wash Plate minimizes risk of cross-contamination

Specifications

Format 8-well Strip
Sample Type E. coli Culture
Efficiency Binding capacity: 20μg
Elution Volume Elution volume: 75–150μL
Final Product Type Sequencing grade plasmid DNA
For Use With (Equipment) Liquid handling instruments
For Use With (Application) Manual and automated processing under vacuum
Includes NucleoSpin Plasmid Filter Strips (12), NucleoSpin Plasmid Binding Strips (12), MN Wash Plate (1), Culture plate with Gas-permeable Foil (1), Rack of Tube Strips (1), RNase A (30mg), Resuspension Buffer A1 (75mL), Lysis Buffer A2 (75mL), Neutralization Buffer A3 (100mL), Wash Buffer AW (100mL), Wash Buffer A4 Concentrate (100mL), Elution Buffer AE (30mL)
No. of Reactions 12 x 8 Preps
Product Type NucleoSpin™ 8 Plasmid Kit
Quantity 1 Kit
Purity A260/A280: 1.70–1.85
Sample Volume 1–5mL
Size <15 kbp
Test Time Preparation time: 45 min. / 6 strips
Yield 4–6μg plasmid per 1mL E. coli culture
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt; guanidine hydrochloride; 2-
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.