missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Virus, Mini kit for viral RNA/DNA from cell-free fluids

Product Code. 12718632
Click to view available options
No. of Reactions:
12 x 8
60 x 8
Unit Size:
480 reactions
96 reactions
This item is not returnable. View return policy

Product Code. 12718632

Brand: Macherey-Nagel™ 740975.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapid isolation of DNA from food and feed in flexible 8‐well strip format.

The NucleoSpin™ Food and NucleoSpin™ Food 8/96 systems guarantee good recovery rates even for small genomic DNA fragments (<1kb) out of processed, complex food matrices, which generally have very low DNA contents as well as poor quality, degraded DNA.
After the food samples have been homogenised, the DNA is extracted with lysis buffers containing chaotropic salts, denaturing agents and detergents. The standard isolation ensures lysis using buffer CF, which was especially developed in cooperation with GEN-IAL, Troisdorf, Germany (patent pending). Lysis mixtures should be cleared by centrifugation or filtration in order to remove contaminants and residual cellular debris. The clear supernatant is then mixed with the binding buffer and ethanol to create conditions for optimal binding to the NucleoSpin™ silica membrane, selected for this purpose due to its unique DNA binding properties. After washing with two different buffers for efficient of potential PCR* inhibitors, DNA can be eluted in low salt buffer or water. Resulting eluates are ready-to-use in all types of subsequent detection methods, especially real-time and basic PCR* technologies and can be used for the identification of GMO-DNA or animal components in food and feed.
Kit components
NucleoSpin™ Food - NucleoSpin™ Food columns, collecting tubes 2mL, buffers, proteinase K. NucleoSpin™ 8 Food - NucleoSpin™ Food binding strips, round-well blocks, MN wash plates, racks with tube strips, cap strips, gas-permeable foil, buffers, proteinase K.NucleoSpin™ 96 Food - NucleoSpin™ 96 Food binding plates, round-well blocks, MN square-well blocks, MN wash plates, racks with tube strips, gas-permeable foil, buffers, proteinase K.
  • Silica membrane technology
  • Available formats: NucleoSpin - mini spin column NucleoSpin 8 - 8 well strip NucleoSpin 96 - 96 well plate
  • Yield: 0.1μg to 10μg
  • Sample material: ≤200μg food/feed
  • Elution volume: 100μL (mini spin column) 100μL to 200μL (8 or 96 well formats)
  • Preparation time: 30min/6 preps (mini spin columns) <60min/8 strips (excluding lysis) <120min/96 well plate (excluding lysis)
  • Manual or automated processing, vacuum/centrifugation
  • No use of organic solvents
Designed for medium to high-throughput isolation of GMO-DNA or animal components in food and feed.

Specifications

Format 8-well Strip
Isolation Technology Silica Membrane Technology
For Use With (Application) For Manual and Automated Isolation of DNA
No. of Reactions 60 x 8
Product Type NucleoSpin™ 8 Food Binding Strip
Test Time 60 min.
Yield 0.1 to 10 μg
compliance-icons compliance-icons compliance-icons
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.