Learn More
Macherey-Nagel™ NucleoSpin™ 8 Blood, 8-well kit for DNA from blood
Description
NucleoSpin™ 8 Blood is designed for the parallel isolation of genomic DNA from whole blood. After lysis of blood cells genomic DNA is selectively bound to a silica membrane while contaminants are washed away effectively. Elution is possible even under vacuum without cross-contamination. Due to its plate design and the possibility to perform the complete procedure under vacuum, NucleoSpin™ 8 Blood can be easily adapted to common liquid handling instruments for fully automated processing without manual interactions. Kit components: NucleoSpin™ Blood binding strips, MN wash plates, MN tube strips, buffers, proteinase K.
Required hardware:
Vacuum processing - the NucleoSpin 8 Blood kits can be used manually with the NucleoVac 96 vacuum manifold. For vacuum processing of the 8-well Strips the reusable Starter Set A, containing Column Holders A and Dummy Strips is required- Silica membrane technology
- Yield: 4μg to 6μg
- Sample material: ≤200μL whole blood, 2 x 106 cultured cells
- Elution volume: 100μL
- Binding capacity: 20μg
- Preparation time: 35min
- Fragment size: 300bp to 30kb Manual or automated isolation of genomic DNA from whole blood or cultured cells.
Specifications
Specifications
| No. of Reactions | 12 x 8 Preps |
| Product Type | 8 Blood |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1