missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 8 Blood Core Kit, 8-well kit for DNA from blood
Description
- Sample Material: Cells, Whole blood
- Sample Amount: 200 μL whole blood (human or animal, fresh or frozen, treated with citrate, EDTA, heparin, or CPDA), 2 x 106 cultured cells
- Fragment Size: 300 bp to 50 kbp
- Binding Capacity: 20 μg
- Preparation Time: 35 min./6 strips
Specifications
Specifications
| Format | 8-well Strip |
| Isolation Technology | Silica Membrane Technology |
| For Use With (Application) | Isolation of DNA |
| Includes | NucleoSpin™ Blood Binding Strips (48 x 8), buffers, Proteinase K |
| No. of Reactions | 384 Preps |
Safety and Handling
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?