Learn More
Macherey-Nagel™ NucleoSnap™ Plasmid Midi Kit
Ultrafast Plasmid Midi Kit for transfection-grade DNA isolation applications, and downstream transfection of sensitive cells and cell injections.
Brand: Macherey-Nagel™ 740494.10
101.81 GBP valid until 2024-12-31
Use promo code "23283" to get your promotional price.
Additional Details : Weight : 0.44000kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Description
- Ultrafast Plasmid Midi Prep due to vacuum processing
- Snap off column
- For vacuum, centrifugation or elution
Specifications
Bacteria, E coli. (50 mL) | |
200 to 500 μL | |
A260/A280: 1.8 to 1.9 | |
<25 kbp |
Bacteria, E. coli | |
Vacuum elution for transfection-grade plasmid DNA isolation and downstream transfection of sensitive cells and cell injections | |
Plasmid DNA | |
250 μg |
Safety and Handling
- NucleoSnap Plasmid Midi
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : RNase; sodium hydroxide solution (diluted < 2 %), 2-propanol