missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSnap cfDNA kit for cell-free DNA from plasma
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
50 reactions
Description
The NucleoVac 24 Vacuum Manifold allows the processing of silica columns for nucleic acid purification by vacuum.
- Snap-off column design for quick processing of large sample volumes
- Compatible with the NucleoVac 24 vacuum manifold (sold separately, MPN 740299
- No need for carrier-RNA
- Process: DNA precipitation—filtration—washing—elution
- Optimized protocol for use with EDTA or Cell-Free DNA BCT™ (Streck) blood draw tubes
- Highly efficient recovery of cell-free DNA from Liquid Biopsies
- No alcohol precipitation necessary
- Typical downstream applications: NGS, qPCR, ddPCR
Specifications
Specifications
| Format | Snap-off Column |
| Isolation Technology | Silica Membrane Technology |
| Sample Type | Plasma, Urine |
| Elution Volume | Elution volume: 20 - 100 μL |
| Final Product Type | Cell-free DNA |
| For Use With (Equipment) | Vacuum (e.g. NucleoVac 24 Vacuum Manifold and vacuum pump) |
| For Use With (Application) | DNA isolation from blood plasma |
| Includes | NucleoSnap DNA Plasma Columns, Collection Tubes, buffers, Proteinase K |
| No. of Reactions | 10 |
| Purity | Fragment size: ≥ 50 bp |
| Show More |
Safety and Handling
Product Identifier
- NucleoSnap DNA Plasma
- Danger
- Flammable liquid Category 2
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- MIXTURE LIST-Contains : potassium hydroxidesolution/proteinaseK, liquid (origin: tritirachium album)/guanidine hydrochlori
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction