missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSEQ™ kit for dye terminator removal

Product Code. p-9281002
Click to view available options
Content And Storage:
10 Preps
250 Preps
50 Preps
Unit Size:
10 reactions
250 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 12768462

Brand: Macherey-Nagel™ 740523.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prefilled single spin columns for dye terminator removal.

  • Contains NucleoSEQ™ columns, collection tubes, and a user manual
  • Designed for fast and effective clean-up of nucleic acids
  • Using gel exclusion in a convenient spin column format allows reliable removal of smaller molecules from nucleic acids
  • Impurities, for example, salts, excess of labels, nucleotides, traces of organic solvents, primers are retained by the column while nucleic acids of interest are recovered with high yield
  • The columns are pre-filled with size exclusion matrix
  • Maximum sample volume to be loaded onto the column: 20 μL
  • Columns with dry gel matrix can be stored at room temperature for 12 months
  • Columns with hydrated gel matrix can be stored at 4°C for 14 days

Specifications

Content And Storage 10 Preps
Description 5 min/prep (without hydration of matrix)
For Use With (Application) Dye terminator removal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.