missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSEQ™ kit for dye terminator removal
Description
- Contains NucleoSEQ™ columns, collection tubes, and a user manual
- Designed for fast and effective clean-up of nucleic acids
- Using gel exclusion in a convenient spin column format allows reliable removal of smaller molecules from nucleic acids
- Impurities, for example, salts, excess of labels, nucleotides, traces of organic solvents, primers are retained by the column while nucleic acids of interest are recovered with high yield
- The columns are pre-filled with size exclusion matrix
- Maximum sample volume to be loaded onto the column: 20 μL
- Columns with dry gel matrix can be stored at room temperature for 12 months
- Columns with hydrated gel matrix can be stored at 4°C for 14 days
Specifications
Specifications
| Content And Storage | 50 Preps |
| Description | 5 min/prep (without hydration of matrix) |
| For Use With (Application) | Dye terminator removal |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?